Search
Search

Stat3| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504646.2 | XP_003504694.1 | 2 - 3 | 348 | 60 | |
| XM_003504647.2 | XP_003504695.1 | 2 - 3 | 349 | 60 | ||
| XM_007645137.1 | XP_007643327.1 | 2 - 3 | 348 | 60 | ||
Set Membership: |
|