Search
Search

Ddx42| Gene Symbol: Ddx42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501868.2 | XP_003501916.1 | 9 - 10 | 1198 | 62 | |
| GenBank mRNA | JI895658.1 | - | 4 - 5 | 555 | 62 | |
| JP045681.1 | - | 4 - 5 | 559 | 62 | ||
Set Membership: |
|