Search
Search

Supv3l1| Gene Symbol: Supv3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499894.2 | XP_003499942.1 | 7 - 8 | 981 | 90 | |
| XM_007640640.1 | XP_007638830.1 | 7 - 8 | 982 | 90 | ||
| GenBank mRNA | JI885688.1 | - | 7 - 8 | 826 | 90 | |
| JP051914.1 | - | 7 - 8 | 825 | 90 | ||
Set Membership: |
|