Search
Search

LOC100772745| Gene Symbol: LOC100772745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505315.2 | XP_003505363.1 | 6 - 7 | 705 | 112 | |
| XM_007645830.1 | XP_007644020.1 | 6 - 7 | 705 | 112 | ||
| XM_007645831.1 | XP_007644021.1 | 6 - 7 | 705 | 112 | ||
| GenBank mRNA | JI874178.1 | - | 3 - 4 | 334 | 112 | |
| JP046700.1 | - | 3 - 4 | 334 | 112 | ||
Set Membership: |
|