Search
Search

Hps1| Gene Symbol: Hps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007617395.1 | XP_007615585.1 | 18 - 19 | 2053 | 62 | |
| XM_007617403.1 | XP_007615593.1 | 18 - 19 | 2061 | 62 | ||
| XM_007617412.1 | XP_007615602.1 | 18 - 19 | 2008 | 62 | ||
Set Membership: |
|