Search
Search

Faf1| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496486.2 | XP_003496534.2 | 17 - 18 | 1643 | 66 | |
| GenBank mRNA | JI867215.1 | - | 7 - 8 | 684 | 66 | |
| JP046271.1 | - | 7 - 8 | 683 | 66 | ||
Set Membership: |
|