Search
Search

Mdh1b| Gene Symbol: Mdh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502117.2 | XP_003502165.1 | 7 - 8 | 1399 | 67 | |
| XM_007642716.1 | XP_007640906.1 | 7 - 8 | 1399 | 67 | ||
| XM_007642718.1 | XP_007640908.1 | 5 - 6 | 1003 | 67 | ||
Set Membership: |
|