Search
Search

Hdac7| Gene Symbol: Hdac7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513062.2 | XP_003513110.2 | 23 - 24 | 2755 | 57 | |
| XM_003513064.2 | XP_003513112.2 | 21 - 22 | 2449 | 57 | ||
| XM_007653425.1 | XP_007651615.1 | 23 - 24 | 2851 | 57 | ||
| XM_007653426.1 | XP_007651616.1 | 24 - 25 | 3042 | 57 | ||
| XM_007653427.1 | XP_007651617.1 | 24 - 25 | 3046 | 57 | ||
| XM_007653428.1 | XP_007651618.1 | 22 - 23 | 2644 | 57 | ||
| XM_007653429.1 | XP_007651619.1 | 22 - 23 | 2623 | 57 | ||
| XM_007653430.1 | XP_007651620.1 | 22 - 23 | 2560 | 57 | ||
Set Membership: |
|