Search
Search

LOC100754516| Gene Symbol: LOC100754516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643800.1 | XP_007641990.1 | 2 - 3 | 665 | 58 | |
| XM_007643801.1 | XP_007641991.1 | 2 - 3 | 665 | 58 | ||
Set Membership: |
|