Search
Search

Kcmf1| Gene Symbol: Kcmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646129.1 | XP_007644319.1 | 6 - 7 | 1020 | 75 | |
| XM_007646130.1 | XP_007644320.1 | 6 - 7 | 987 | 75 | ||
| XM_007646131.1 | XP_007644321.1 | 6 - 7 | 960 | 75 | ||
| XM_007646132.1 | XP_007644322.1 | 6 - 7 | 948 | 75 | ||
| XM_007646133.1 | XP_007644323.1 | 5 - 6 | 889 | 75 | ||
| GenBank mRNA | JP051671.1 | - | 5 - 6 | 917 | 75 | |
Set Membership: |
|