Search
Search

Srpk3| Gene Symbol: Srpk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506536.2 | XP_003506584.1 | 9 - 10 | 903 | 56 | |
| XM_007647047.1 | XP_007645237.1 | 9 - 10 | 903 | 56 | ||
Set Membership: |
|