Search
Search

Pak3| Gene Symbol: Pak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649122.1 | XP_007647312.1 | 12 - 13 | 1322 | 91 | |
| XM_007649123.1 | XP_007647313.1 | 13 - 14 | 1433 | 91 | ||
| XM_007649124.1 | XP_007647314.1 | 13 - 14 | 1413 | 91 | ||
| XM_007649125.1 | XP_007647315.1 | 12 - 13 | 1328 | 91 | ||
| GenBank mRNA | JP056069.1 | - | 11 - 12 | 1427 | 91 | |
Set Membership: |
|