Search
Search

LOC100772575| Gene Symbol: LOC100772575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648935.1 | XP_007647125.1 | 2 - 3 | 483 | 85 | |
| GenBank mRNA | JI865621.1 | - | 2 - 3 | 307 | 85 | |
Set Membership: |
|