Search
Search

Pja2| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499365.2 | XP_003499413.1 | 6 - 7 | 1873 | 73 | |
| XM_003499366.2 | XP_003499414.1 | 5 - 6 | 1687 | 73 | ||
| GenBank mRNA | JI893107.1 | - | 2 - 3 | 294 | 73 | |
| JP046985.1 | - | 2 - 3 | 272 | 73 | ||
Set Membership: |
|