Search
Search

Supt20h| Gene Symbol: Supt20h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652509.1 | XP_007650699.1 | 15 - 16 | 1502 | 67 | |
| XM_007652510.1 | XP_007650700.1 | 15 - 16 | 1503 | 67 | ||
| GenBank mRNA | JI877441.1 | - | 4 - 5 | 67 | ||
Set Membership: |
|