Search
Search

Bola3| Gene Symbol: Bola3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510030.2 | XP_003510078.1 | 1 - 2 | 127 | 87 | |
| GenBank mRNA | JI893933.1 | - | 1 - 2 | 128 | 87 | |
| JP046833.1 | - | 1 - 2 | 128 | 87 | ||