Search
Search

Ablim1| Gene Symbol: Ablim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653778.1 | XP_007651968.1 | 15 - 16 | 1185 | 67 | |
| XM_007653779.1 | XP_007651969.1 | 18 - 19 | 1480 | 67 | ||
Set Membership: |
|