Search
Search

LOC100753013| Gene Symbol: LOC100753013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499991.2 | XP_003500039.1 | 2 - 3 | 298 | 78 | |
| XM_007640725.1 | XP_007638915.1 | 2 - 3 | 232 | 78 | ||
| XM_007640727.1 | XP_007638917.1 | 2 - 3 | 330 | 78 | ||
| XM_007640728.1 | XP_007638918.1 | 2 - 3 | 291 | 78 | ||
| XM_007640729.1 | XP_007638919.1 | 2 - 3 | 155 | 78 | ||
| GenBank mRNA | JI892364.1 | - | 1 - 2 | 136 | 78 | |