Search
Search

Pkn3| Gene Symbol: Pkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514661.2 | XP_003514709.1 | 13 - 14 | 1984 | 55 | |
| XM_007654728.1 | XP_007652918.1 | 13 - 14 | 1717 | 55 | ||
| XM_007654729.1 | XP_007652919.1 | 13 - 14 | 1780 | 55 | ||
| XM_007654730.1 | XP_007652920.1 | 13 - 14 | 1813 | 55 | ||
Set Membership: |
|