Search
Search

Kiaa0556| Gene Symbol: Kiaa0556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650723.1 | XP_007648913.1 | 18 - 19 | 3644 | 56 | |
| XM_007650724.1 | XP_007648914.1 | 19 - 20 | 3714 | 56 | ||
| XM_007650725.1 | XP_007648915.1 | 17 - 18 | 3612 | 56 | ||
| XM_007650726.1 | XP_007648916.1 | 17 - 18 | 3566 | 56 | ||
| XM_007650727.1 | XP_007648917.1 | 17 - 18 | 3529 | 56 | ||
| XM_007650728.1 | XP_007648918.1 | 18 - 19 | 3775 | 56 | ||
Set Membership: |
|