Search
Search

Wwp1| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652180.1 | XP_007650370.1 | 8 - 9 | 1109 | 72 | |
| GenBank mRNA | JI892705.1 | - | 1 - 2 | 302 | 72 | |
| JP047707.1 | - | 1 - 2 | 302 | 72 | ||
Set Membership: |
|