Search
Search

ATP2C1| Gene Symbol: ATP2C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051409.1 | XP_017906898.1 | 13 - 14 | 1524 | 92 | |
| XM_018051417.1 | XP_017906906.1 | 15 - 16 | 1471 | 92 | ||
| XM_018051427.1 | XP_017906916.1 | 15 - 16 | 1312 | 92 | ||
| XM_018051432.1 | XP_017906921.1 | 14 - 15 | 1407 | 92 | ||
| XM_018051436.1 | XP_017906925.1 | 13 - 14 | 1282 | 92 | ||
| GenBank mRNA | JO426167.1 | - | 14 - 15 | 1951 | 92 | |
Set Membership: |
|