Search
Search
| Gene Symbol: EXTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051858.1 | XP_017907347.1 | 5 - 6 | 3094 | 80 | |
| XM_018051859.1 | XP_017907348.1 | 6 - 7 | 3208 | 80 | ||
| XM_018051860.1 | XP_017907349.1 | 5 - 6 | 3101 | 80 | ||
Set Membership: |
|