Search
Search

PLEKHA3| Gene Symbol: PLEKHA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065208.1 | XP_017920697.1 | 5 - 6 | 1521 | 68 | |
| GenBank mRNA | JT677359.1 | - | 5 - 6 | 1011 | 68 | |
Set Membership: |
|