Search
Search
| Gene Symbol: SETD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066690.1 | XP_017922179.1 | 4 - 5 | 1134 | 60 | |
| XM_018066691.1 | XP_017922180.1 | 8 - 9 | 1145 | 60 | ||
| XM_018066692.1 | XP_017922181.1 | 7 - 8 | 920 | 60 | ||
| XM_018066693.1 | XP_017922182.1 | 6 - 7 | 727 | 60 | ||
| XM_018066694.1 | XP_017922183.1 | 4 - 5 | 1132 | 60 | ||
| XM_018066695.1 | XP_017922184.1 | 4 - 5 | 1132 | 60 | ||
| XM_018066696.1 | XP_017922185.1 | 4 - 5 | 1077 | 60 | ||
| XM_018066697.1 | XP_017922186.1 | 6 - 7 | 727 | 60 | ||
| XM_018066698.1 | XP_017922187.1 | 6 - 7 | 827 | 60 | ||
| XM_018066699.1 | XP_017922188.1 | 7 - 8 | 1052 | 60 | ||
| GenBank mRNA | JO402634.2 | - | 18 - 19 | 4362 | 60 | |
| JO402635.2 | - | 18 - 19 | 4362 | 60 | ||
Set Membership: |
|