Search
Search
| Gene Symbol: GIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061109.1 | XP_017916598.1 | 10 - 11 | 1051 | 64 | |
| XM_018061110.1 | XP_017916599.1 | 10 - 11 | 1023 | 64 | ||
| XM_018061111.1 | XP_017916600.1 | 10 - 11 | 1051 | 64 | ||
| XM_018061112.1 | XP_017916601.1 | 10 - 11 | 1051 | 64 | ||
| XM_018061113.1 | XP_017916602.1 | 10 - 11 | 1051 | 64 | ||
| XM_018061114.1 | XP_017916603.1 | 10 - 11 | 1048 | 64 | ||
| XM_018061115.1 | XP_017916604.1 | 10 - 11 | 1051 | 64 | ||
| XM_018061116.1 | XP_017916605.1 | 10 - 11 | 1051 | 64 | ||
Set Membership: |
|