Search
Search
| Gene Symbol: HMOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285567.1 | NP_001272496.1 | 3 - 4 | 638 | 78 | |
| GenBank mRNA | JF731041.1 | - | 3 - 4 | 638 | 78 | |
| JO580392.1 | - | 3 - 4 | 808 | 78 | ||
| KA334415.1 | - | 3 - 4 | 790 | 78 | ||
Set Membership: |
|