Search
Search

MYO18A| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064334.1 | XP_017919823.1 | 32 - 33 | 4830 | 58 | |
| XM_018064335.1 | XP_017919824.1 | 31 - 32 | 4770 | 58 | ||
| XM_018064336.1 | XP_017919825.1 | 30 - 31 | 4749 | 58 | ||
| XM_018064337.1 | XP_017919826.1 | 30 - 31 | 4734 | 58 | ||
| XM_018064338.1 | XP_017919827.1 | 29 - 30 | 4713 | 58 | ||
| XM_018064339.1 | XP_017919828.1 | 34 - 35 | 5553 | 58 | ||
| XM_018064340.1 | XP_017919829.1 | 29 - 30 | 3618 | 58 | ||
| XM_018064341.1 | XP_017919830.1 | 29 - 30 | 4712 | 58 | ||
| XM_018064342.1 | XP_017919831.1 | 29 - 30 | 4712 | 58 | ||
| XM_018064343.1 | XP_017919832.1 | 30 - 31 | 4749 | 58 | ||
| GenBank mRNA | JO424276.1 | - | 13 - 14 | 3028 | 58 | |
Set Membership: |
|