Search
Search

ARHGAP9| Gene Symbol: ARHGAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048025.1 | XP_017903514.1 | 12 - 13 | 1881 | 56 | |
| XM_018048026.1 | XP_017903515.1 | 11 - 12 | 1187 | 56 | ||
| GenBank mRNA | JO418274.2 | - | 3 - 4 | 252 | 56 | |
Set Membership: |
|