Search
Search
| Gene Symbol: CSDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681114.3 | XP_005681171.1 | 2 - 3 | 641 | 83 | |
| GenBank mRNA | JO632487.2 | - | 2 - 3 | 444 | 83 | |
| KA323684.1 | - | 2 - 3 | 443 | 83 | ||
Set Membership: |
|