Search
Search
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683172.3 | XP_005683229.1 | 3 - 4 | 575 | 84 | |
| XM_005683173.3 | XP_005683230.1 | 3 - 4 | 556 | 84 | ||
| GenBank mRNA | JO580974.1 | - | 3 - 4 | 620 | 84 | |
Set Membership: |
|