Search
Search
| Gene Symbol: UBTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693926.3 | XP_005693983.1 | 15 - 16 | 1777 | 61 | |
| XM_005693927.3 | XP_005693984.1 | 14 - 15 | 1666 | 61 | ||
| XM_018065218.1 | XP_017920707.1 | 15 - 16 | 1794 | 61 | ||
| XM_018065219.1 | XP_017920708.1 | 15 - 16 | 2081 | 61 | ||
| XM_018065220.1 | XP_017920709.1 | 15 - 16 | 1770 | 61 | ||
| XM_018065221.1 | XP_017920710.1 | 14 - 15 | 1665 | 61 | ||
| GenBank mRNA | JO646262.2 | - | 2 - 3 | 288 | 61 | |
Set Membership: |
|