Search
Search

POLR3E| Gene Symbol: POLR3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040861.1 | XP_017896350.1 | 9 - 10 | 828 | 56 | |
| XM_018040862.1 | XP_017896351.1 | 10 - 11 | 932 | 56 | ||
| GenBank mRNA | JO618619.2 | - | 12 - 13 | 1719 | 56 | |
| JT670884.1 | - | 9 - 10 | 719 | 56 | ||
Set Membership: |
|