Search
Search

SLC29A3| Gene Symbol: SLC29A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042501.1 | XP_017897990.1 | 3 - 4 | 465 | 84 | |
| XM_018042502.1 | XP_017897991.1 | 4 - 5 | 514 | 84 | ||
| GenBank mRNA | KA315623.1 | - | 3 - 4 | 498 | 84 | |
Set Membership: |
|