Search
Search
| Gene Symbol: CMSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058548.1 | XP_017914037.1 | 2 - 3 | 309 | 75 | |
| XM_018058605.1 | XP_017914094.1 | 2 - 3 | 114 | 75 | ||
| GenBank mRNA | JO377716.2 | - | 6 - 7 | 503 | 75 | |
| JO416957.2 | - | 6 - 7 | 524 | 75 | ||
| JO443482.2 | - | 2 - 3 | 277 | 75 | ||
| JO515698.2 | - | 6 - 7 | 542 | 75 | ||
| JO516937.2 | - | 6 - 7 | 542 | 75 | ||
| JO549753.2 | - | 2 - 3 | 217 | 75 | ||
| JO591971.2 | - | 8 - 9 | 866 | 75 | ||
| JO591972.2 | - | 8 - 9 | 866 | 75 | ||
| JT705535.1 | - | 1 - 2 | 91 | 75 | ||
| KA318277.1 | - | 7 - 8 | 587 | 75 | ||
Set Membership: |
|