Search
Search
| Gene Symbol: WRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042048.1 | XP_017897537.1 | 5 - 6 | 732 | 65 | |
| XM_018042049.1 | XP_017897538.1 | 5 - 6 | 732 | 65 | ||
| XM_018042050.1 | XP_017897539.1 | 5 - 6 | 732 | 65 | ||
| XM_018042051.1 | XP_017897540.1 | 5 - 6 | 732 | 65 | ||
Set Membership: |
|