Search
Search
| Gene Symbol: KLKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698743.1 | XP_005698800.1 | 6 - 7 | 688 | 58 | |
| XM_013975393.1 | XP_013830847.1 | 6 - 7 | 688 | 58 | ||
| GenBank mRNA | JO420801.2 | - | 6 - 7 | 680 | 58 | |
Set Membership: |
|