Search
Search
| Gene Symbol: POLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699889.2 | XP_005699946.2 | 7 - 8 | 1129 | 59 | |
| XM_013976039.2 | XP_013831493.1 | 7 - 8 | 1129 | 59 | ||
| XM_013976040.2 | XP_013831494.1 | 7 - 8 | 1129 | 59 | ||
| GenBank mRNA | JO604200.2 | - | 7 - 8 | 1095 | 59 | |
Set Membership: |
|