Search
Search

PTDSS2| Gene Symbol: PTDSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043069.1 | XP_017898558.1 | 5 - 6 | 724 | 66 | |
| GenBank mRNA | JO352011.2 | - | 4 - 5 | 402 | 66 | |
| JO543810.2 | - | 4 - 5 | 379 | 66 | ||
Set Membership: |
|