Search
Search

ZNF687| Gene Symbol: ZNF687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046070.1 | XP_017901559.1 | 2 | 1601 | 50 | |
| XM_018046071.1 | XP_017901560.1 | 2 | 1515 | 50 | ||
| XM_018046072.1 | XP_017901561.1 | 2 | 1610 | 50 | ||
Set Membership: |
|