Search
Search

COL4A3| Gene Symbol: COL4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058855.1 | XP_017914344.1 | 18 - 19 | 1197 | 76 | |
| GenBank mRNA | JT672045.1 | - | 20 - 21 | 1474 | 76 | |
Set Membership: |
|