Search
Search
| Gene Symbol: COQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964761.2 | XP_013820215.2 | 6 - 7 | 986 | 70 | |
| GenBank mRNA | JO433330.2 | - | 6 - 7 | 982 | 70 | |
| JO613622.2 | - | 1 - 2 | 648 | 70 | ||
Set Membership: |
|