Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052407.1 | XP_017907896.1 | 2 - 3 | 265 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |