Search
Search

NCKAP1| Gene Symbol: NCKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065461.1 | XP_017920950.1 | 12 - 13 | 1124 | 69 | |
| XM_018065462.1 | XP_017920951.1 | 11 - 12 | 1113 | 69 | ||
Set Membership: |
|