Search
Search

TMEM65| Gene Symbol: TMEM65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058546.1 | XP_017914035.1 | 6 - 7 | 1104 | 76 | |
| XM_018058547.1 | XP_017914036.1 | 5 - 6 | 1020 | 76 | ||
Set Membership: |
|