Search
Search
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049716.1 | XP_017905205.1 | 13 - 14 | 2133 | 54 | |
| XM_018049717.1 | XP_017905206.1 | 13 - 14 | 1905 | 54 | ||
| XM_018049718.1 | XP_017905207.1 | 13 - 14 | 1905 | 54 | ||
| XM_018049719.1 | XP_017905208.1 | 13 - 14 | 1905 | 54 | ||
Set Membership: |
|