Search
Search
| Gene Symbol: RCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696367.3 | XP_005696424.1 | 2 - 3 | 603 | 92 | |
| XM_018039084.1 | XP_017894573.1 | 3 - 4 | 1205 | 92 | ||
| XM_018039085.1 | XP_017894574.1 | 3 - 4 | 426 | 92 | ||
Set Membership: |
|