Search
Search
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040708.1 | XP_017896197.1 | 14 - 15 | 2357 | 60 | |
| GenBank mRNA | JO428438.1 | - | 2 - 3 | 1830 | 60 | |
| JO638095.2 | - | 2 - 3 | 1862 | 60 | ||
Set Membership: |
|