Search
Search
| Gene Symbol: PTPN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680926.2 | XP_005680983.1 | 5 - 6 | 942 | 64 | |
| XM_018048543.1 | XP_017904032.1 | 5 - 6 | 886 | 64 | ||
| GenBank mRNA | KA324698.1 | - | 5 - 6 | 919 | 64 | |
Set Membership: |
|