Search
Search
| Gene Symbol: PLS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965948.2 | XP_013821402.1 | 12 - 13 | 1423 | 72 | |
| XM_018049572.1 | XP_017905061.1 | 12 - 13 | 1555 | 72 | ||
| XM_018049577.1 | XP_017905066.1 | 12 - 13 | 1492 | 72 | ||
| GenBank mRNA | JO432308.2 | - | 4 - 5 | 765 | 72 | |
Set Membership: |
|